missing translation for 'onlineSavingsMsg'
Learn More

1-(pyrid-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride, 97%, Thermo Scientific™

Product Code. 10123044
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10123044 5 g Amber Glass Bottle 5g
10575944 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123044 Supplier Thermo Scientific Maybridge Supplier No. CC69851DE
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 946409-40-1
Molecular Formula C12H18Cl2N2O2
Molecular Weight (g/mol) 293.188
MDL Number MFCD06801260
InChI Key FLINLLCJYJQIIY-UHFFFAOYSA-N
Synonym 1-pyridin-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride, 1-pyrid-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride, 1-pyridin-3-yl methyl piperidine-4-carboxylic acid dihydrochloride, 1-pyridin-3-yl methyl piperidine-4-carboxylic acid-hydrogen chloride 1/2
PubChem CID 42935493
IUPAC Name 1-(pyridin-3-ylmethyl)piperidine-4-carboxylic acid;dihydrochloride
SMILES C1CN(CCC1C(=O)O)CC2=CN=CC=C2.Cl.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 293.19
Percent Purity 95%
Chemical Name or Material 1-(Pyrid-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.