missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrimidin-2-yl-piperidine-4-carboxylic acid, Thermo Scientific™

Product Code. 10401474
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10401474 5 g 5g
10001174 1 g 1g
10546484 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10401474 Supplier Thermo Scientific Maybridge Supplier No. CC20701DE
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 303144-44-7
Molecular Formula C10H12N3O2
Molecular Weight (g/mol) 206.23
MDL Number MFCD00664474
InChI Key DYVXURZASBPYFR-UHFFFAOYSA-M
Synonym 1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl
PubChem CID 2767267
IUPAC Name 1-pyrimidin-2-ylpiperidine-4-carboxylic acid
SMILES [O-]C(=O)C1CCN(CC1)C1=NC=CC=N1

Specifications

Packaging Amber glass bottle
Quantity 5 g
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material 1-Pyrimidin-2-yl-piperidine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.