missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrrolidino-1-cyclohexene, 95%

Product Code. 10552701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10552701 25 g Glass bottle 25g
10307581 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10552701 Supplier Thermo Scientific Acros Supplier No. 334830250
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1125-99-1 | C10H17N | 151.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-99-1
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.25
MDL Number MFCD00003163
InChI Key KTZNVZJECQAMBV-UHFFFAOYSA-N
Synonym 1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine
PubChem CID 70768
IUPAC Name 1-(cyclohexen-1-yl)pyrrolidine
SMILES C1CCC(=CC1)N2CCCC2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 0.9400g/mL
Boiling Point 114°C to 115°C (15.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5210 to 1.523
Quantity 25 g
Specific Gravity 0.94
Formula Weight 151.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Pyrrolidino-1-cyclohexene, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Pyrrolidino-1-cyclohexene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.