missing translation for 'onlineSavingsMsg'
Learn More

1-S-Octyl-β-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals

Product Code. 10137653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137653 2.5 g Glass bottle 2.5g
10731361 500 mg Glass bottle 500mg
10239643 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10137653 Supplier Thermo Scientific Acros Supplier No. 315000025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85618-21-9
Molecular Formula C14H28O5S
Molecular Weight (g/mol) 308.44
MDL Number MFCD00012189
InChI Key CGVLVOOFCGWBCS-DGTMBMJNSA-N
Synonym octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
PubChem CID 53720324
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
SMILES CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 2.5 g
Specific Rotation Condition − 46.00 (20.00°C c=1,CH3OH)
Specific Rotation -46
Solubility Information Solubility in water: soluble
Formula Weight 308.44
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-S-Octyl-β-D-thioglucopyranoside, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.