missing translation for 'onlineSavingsMsg'
Learn More

uvex 1 Sport White NC Shoe O1 FO SRC, Width 11

Product Code. 17893180 Shop All Uvex Products
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17893180 50 1 pair
17843170 35 1 pair
17853170 36 1 pair
17863170 37 1 pair
17873170 38 1 pair
17883170 39 1 pair
17893170 40 1 pair
17803180 41 1 pair
17813180 42 1 pair
17823180 43 1 pair
17833180 44 1 pair
17843180 45 1 pair
17853180 46 1 pair
17863180 47 1 pair
17873180 48 1 pair
17883180 49 1 pair
17803190 51 1 pair
17813190 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 17893180 Supplier uvex Supplier No. 6570850
  • ONLINE EXCLUSIVE PRICE
    131.00 €
    122.32 € / 1 pairs
    Save 8.68 €
    6% Off

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With a lightweight design, high-tech materials, and ergonomically designed outsole, the Uvex 1 Sport White NC Shoe is the ideal companion for the working day and beyond. The hydrophobic upper ensures optimized climate control, and reliably protects the wearer against liquids and other contaminants.

  • Modern, ultra-lightweight and flexible O1 occupational shoe without protective toe cap
  • Washable at up to 30°C
  • Made from synthetic materials, so suitable for people allergic to chrome
  • Elastic laces with quick release for individual adjustment
  • All sole materials are free of silicone, plasticizers and other substances that interfere with wetting agents
  • New, innovative uvex i-PUREnrj sole technology provides outstanding shock-absorption properties at the forefoot and heel and excellent rebound energy over the entire midsole
  • Meets the ESD specification with volume resistance less than 35 Megohms
  • Ergonomically designed outsole made from dual density polyurethane with excellent slip resistance
  • Outstanding wearer comfort thanks to a newly developed last and climate-optimized, breathable materials
  • Virtually seam-free construction with high-tech materials to eliminate pressure points
  • Removable anti-static comfortable insole with a moisture transport system and additional shock absorption in the forefoot and heel
  • Soft padding on collar and tongue
  • Made partly from recycled materials in order to conserve resources
  • Free from any of the harmful substances listed on our banned hazardous substances list
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Textile
Size 50
Toe Material Plain Toe
Certifications/Compliance (Europe) EN ISO 20347:2012 O1 FO SRC EN ISO 20347:2022 O1 FO SC SR
Gender Unisex
Fit Standard Width
Latex Free Yes
Ankle Style Trainers
Waterproof No
Product Type ESD Safety Shoe
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.