Learn More
Description
The Thermo Scientific™ 1-Step Human High-Yield In Vitro Translation (IVT) Kits with continuous-feed device enables the expression of functional proteins with 10 to 100 times greater yield per mL than other eukaryotic IVT translation systems.
Highlights:
High expression – up to 750μg/mL of expressed protein
Reproducibility – low variability between experiments
Fast – express high levels of protein with an overnight incubation
Functional – obtain more active protein than with bacterial protein expression systems
1-Step Protein Expression:
Streamlined reaction – requires only five components
Easy-to-use – transcription and translation is performed in one reaction step
Adapt for labeling – amenable to incorporation of heavy or unnatural amino acids
Includes:
Complete kits contain cell lysate, reaction mix, accessory proteins, 5X dialysis buffer, control DNA, cloning vector, mini dialysis devices and centrifuge tubes
Requires:
30°C incubator or water bath
Recommended for:
Production of recombinant proteins for structural analysis; Generate active proteins for protein interaction studies; Perform rapid expression and characterization of mutant proteins; Small-scale production of virual particles; Incorporation of unnatural amino acids or toxic protein production
Spezifikation
Spezifikation
| Type | 1-step Human High-yield Mini IVT Kit |
| Sufficient For | 10 Reactions of 100 μL Each |
| Quantity | 10 x 100 μL |
| Content And Storage | Sufficient For: 10 reactions of 100 μL each • HeLa Lysate, 500 μL • Accessory Proteins, 100 μL • Reaction Mix, 200 μL • 5X Dialysis Buffer, 3 mL • Positive Control DNA (pCFE-GFP), 10 μg • pT7CFE1-NHis-GST-CHA, 10 μg • Microdialysis Device, 10 each • Nuclease-free Water, 3 x 5 mL Store micro-dialysis devices and nuclease-free water at room temperature. Store all other components at -80°C. |
| Expression System | Cell-Free |
| Expressed Protein Type | Insoluble Protein |
| Product Line | 1-Step |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.