missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step™ Ultra TMB-Blotting Solution

Product Code. 13473869 Shop All Thermo Scientific Products
Encompass_Preferred
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 13473869

Brand: Thermo Scientific™ 37574

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.

Thermo Scientific™ 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.

Additional Information and Online Ordering

Thermo Scientific 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting.

This precipitating, colorimetric Western blot substrate for HRP provides high sensitivity, increased signal-to-noise ratio and low background compared to many other chromogenic substrates. The blotting solution contains soluble TMB (3,3',5,5'-tetramethylbenzidine), which reacts very quickly with horseradish peroxidase enzyme to produce an insoluble dark blue precipitate that does not fade or flake. The substrate is compatible with both nitrocellulose and PVDF membranes. The blotting solution is supplied ready to use with no mixing required.

Features of 1-Step Ultra TMB-Blotting Solution:
Fast—protein bands visible in less than one minute
Fade-resistant—protein bands stable after membrane drying
Sensitive—detection limit similar to Pierce ECL Western Blotting Substrate
Chromogenic—no special equipment needed for visualization; produces dark blue bands
Ready to use—no organic solvents are required to dissolve; no dilution necessary for use

Applications:
• Chromogenic Western blot detection
• Nitrocellulose membrane blots
• PVDF membrane blots

Specifications

Product Type Substrate
Detection Method Colorimetric
Form Liquid
Membrane Compatibility Nitrocellulose, PVDF
Product Line 1-Step
Quantity 250 mL
Substrate TMB
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.