missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-2-indolinecarboxylic acid, 90%, Thermo Scientific™

Product Code. 10238722
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10238722 1 g 1g
10249052 250 mg 250mg
10628160 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10238722 Supplier Thermo Scientific Maybridge Supplier No. MO01075DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 133851-52-2
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD02682411
InChI Key QONNUMLEACJFME-UHFFFAOYSA-N
Synonym 1-tert-butoxycarbonyl-2-indolinecarboxylic acid, n-boc-indoline-2-carboxylic acid, 1-boc-indoline-2-carboxylic acid, 1-tert-butoxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydroindole-2-carboxylic acid, 1-n-boc-indoline-2-carboxylic acid, 1-2-methylpropan-2-yl oxycarbonyl-2,3-dihydroindole-2-carboxylic acid, indoline-2-carboxylic acid, n-boc protected, 1-tert-butoxycarbonyl indoline-2-carboxylicacid
PubChem CID 2794663
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)C(=O)O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 263.29
Percent Purity 90%
Chemical Name or Material 1-(Tert-butoxycarbonyl)-2-indolinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.