missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-2-indolinecarboxylic acid, 90%, Thermo Scientific™

Product Code. 10249052
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10249052 250 mg 250mg
10238722 1 g 1g
10628160 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10249052 Supplier Thermo Scientific Maybridge Supplier No. MO01075CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 133851-52-2
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD02682411
InChI Key QONNUMLEACJFME-UHFFFAOYSA-N
Synonym 1-tert-butoxycarbonyl-2-indolinecarboxylic acid, n-boc-indoline-2-carboxylic acid, 1-boc-indoline-2-carboxylic acid, 1-tert-butoxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydroindole-2-carboxylic acid, 1-n-boc-indoline-2-carboxylic acid, 1-2-methylpropan-2-yl oxycarbonyl-2,3-dihydroindole-2-carboxylic acid, indoline-2-carboxylic acid, n-boc protected, 1-tert-butoxycarbonyl indoline-2-carboxylicacid
PubChem CID 2794663
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)C(=O)O

Tekniske data

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 263.29
Percent Purity 90%
Chemical Name or Material 1-(Tert-butoxycarbonyl)-2-indolinecarboxylic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.