missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyl) 2-methyl 1,2-indolinedicarboxylate, 97%, Thermo Scientific™

Product Code. 10587274
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10587274 5 g 5g
10424333 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10587274 Supplier Thermo Scientific Maybridge Supplier No. MO01034DE
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 186704-03-0
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
MDL Number MFCD06200867
InChI Key JWTYDRKOSWALHD-UHFFFAOYSA-N
Synonym 1-tert-butyl 2-methyl 1,2-indolinedicarboxylate, 1-tert-butyl 2-methyl indoline-1,2-dicarboxylate, 1-tert-butyl-2-methylindoline-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2,3-dihydroindole-1,2-dicarboxylate, 1-boc-indolin-2-carboxylic acid methyl ester, 1-tert-butyl-2-methyl-1,2-indoline dicarboxylate, 1-tert-butoxycarbonyl indoline-2-carboxylic acid methyl ester
PubChem CID 2794661
IUPAC Name 1-O-tert-butyl 2-O-methyl 2,3-dihydroindole-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)C(=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 277.32
Percent Purity 97%
Chemical Name or Material 1-(Tert-butyl) 2-methyl 1, 2-indolinedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.