missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10633455
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10633455 5 g 5g
10311353 250 mg 250mg
10615753 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10633455 Supplier Thermo Scientific Maybridge Supplier No. CD07394DE
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 647824-51-9
Molecular Formula C10H16N2O
Molecular Weight (g/mol) 180.251
MDL Number MFCD00120293
InChI Key WQHXOHHEPNBXMF-UHFFFAOYSA-N
Synonym 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carbaldehyde, 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carboxaldehyde, buttpark 99\12-15, 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde, acmc-20ap9c
PubChem CID 2776936
IUPAC Name 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde
SMILES CC1=C(C(=NN1C(C)(C)C)C)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material 1-(Tert-butyl)-3, 5-dimethyl-1H-pyrazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.