missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-3,5-dimethylbenzene, TRC

Product Code. 30338821
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338821 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338821 Supplier Toronto Research Chemicals Supplier No. TRCB699318100G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 5-tert-Butyl-m-Xylene
CAS 98-19-1
Molecular Formula C12 H18
Quantity 100 g
Synonym 1-(1,1-Dimethylethyl)-3,5-dimethylbenzene, Xylometazoline Hydrochloride Imp. D (EP)
InChI Formula InChI=1S/C12H18/c1-9-6-10(2)8-11(7-9)12(3,4)5/h6-8H,1-5H3
SMILES Cc1cc(C)cc(c1)C(C)(C)C
IUPAC Name 1-tert-butyl-3,5-dimethylbenzene
Molecular Weight (g/mol) 162.27
Formula Weight 162.1409
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.