missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl 6-methyl 3-iodo-1H-indole-1,6-dicarboxylate, ≥97%, Thermo Scientific™

Product Code. 10442722
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10442722 1 g Amber Glass Bottle 1g
10340213 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10442722 Supplier Thermo Scientific Maybridge Supplier No. MO07419DA
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 850374-94-6
Molecular Formula C15H16INO4
Molecular Weight (g/mol) 401.2
MDL Number MFCD06797499
InChI Key KFPCBJZNHXTGHX-UHFFFAOYSA-N
Synonym 1-tert-butyl 6-methyl 3-iodo-1h-indole-1,6-dicarboxylate, 1-tert-butyl 6-methyl-3-iodo-1h-indole-1,6-dicarboxylate, 1-tert-butyl 6-methyl 3-iodoindole-1,6-dicarboxylate, 1-tert-butyl-6-methyl-3-iodo-1h-indole-1,6-dicarboxylate, methyl 3-iodo-1h-indole-6-carboxylate,n-boc protected, 1-tert-butyl-6-methyl 3-iodo-1h-indole-1,6-dicarboxylate
PubChem CID 7127827
IUPAC Name 1-O-tert-butyl 6-O-methyl 3-iodoindole-1,6-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C=C(C2=C1C=C(C=C2)C(=O)OC)I

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 401.19
Percent Purity 97%
Chemical Name or Material 1-(tert-Butyl)-6-methyl-3-iodo-1H-indole-1, 6-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.