missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyldimethylsilyl)imidazole, 97%

Product Code. 11364198
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364198 25 g 25g
11354198 1 g 1g
11344198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364198 Supplier Thermo Scientific Alfa Aesar Supplier No. 041462.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54925-64-3 | C9H18N2Si | 182.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54925-64-3
Molecular Formula C9H18N2Si
Molecular Weight (g/mol) 182.34
MDL Number MFCD00011682
InChI Key VUENSYJCBOSTCS-UHFFFAOYSA-N
Synonym 1-tert-butyldimethylsilyl-1h-imidazole, 1-tert-butyldimethylsilyl imidazole, tert-butyldimethylsilylimidazole, n-tert-butyldimethylsilylimidazole, t-butyldimethylsilylimidazole, 1-t-butyldimethylsilyl imidazole, 1h-imidazole, 1-1,1-dimethylethyl dimethylsilyl, tbdmsim, imidazole, tbdms derivative, 1-tert-butyldimethylsilylimidazole
PubChem CID 171385
IUPAC Name tert-butyl-imidazol-1-yl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)N1C=CN=C1

Specifications

Density 0.94 g/mL
Boiling Point 247°C to 250°C
Flash Point 99°C (210°F)
Refractive Index 1.4805
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 182.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(tert-Butyldimethylsilyl)imidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.