missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecylphosphonic acid, 98%

Product Code. 11408127
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408127 1 g 1g
11418127 5 g 5g
11428127 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408127 Supplier Thermo Scientific Alfa Aesar Supplier No. A17511.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4671-75-4 | C14H31O3P | 278.373 g/mol

1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.

Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4671-75-4
Molecular Formula C14H31O3P
Molecular Weight (g/mol) 278.373
MDL Number MFCD00015834
InChI Key BVQJQTMSTANITJ-UHFFFAOYSA-N
Synonym phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate
PubChem CID 78401
IUPAC Name tetradecylphosphonic acid
SMILES CCCCCCCCCCCCCCP(=O)(O)O

Specifications

Quantity 1 g
Solubility Information Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.
Formula Weight 278.37
Percent Purity 98%
Chemical Name or Material 1-Tetradecylphosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.