missing translation for 'onlineSavingsMsg'
Learn More

1-(Tetrahydro-2-furoyl)-piperazine, TRC

Product Code. 30421152
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Product Code. Quantity unitSize
30421152 500 mg 500mg
30412174 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 30421152

Brand: Toronto Research Chemicals TRCT293475500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[[(2RS)-tetrahydrofuran-2-yl]carbonyl]piperazine
CAS 63074-07-7
Molecular Formula C9 H16 N2 O2
Quantity 500 mg
Synonym 1-[[(2RS)-Tetrahydrofuran-2-yl]carbonyl]piperazine, Methanone','1-piperazinyl(tetrahydro-2-furanyl)-, Piperazine','1-[(tetrahydro-2-furanyl)carbonyl]- (9CI), 1-Piperazinyl(tetrahydro-2-furanyl)methanone, (Piperazin-1-yl)(tetrahydrofuran-2-yl)methanone, 1-(Tetrahydro-2-furoyl)piperazine, 1-(Tetrahydrofuran-2-carbonyl)piperazine, 1-(Tetrahydrofuran-2-ylcarbonyl)piperazine, 1-[(2,3,4,5-Tetrahydrofuran-2-yl)carbonyl]piperazine, 4-[(Tetrahydrofuran-2-yl)carbonyl]piperazine, N-(Tetrahydro-2-furoyl)piperazine
InChI Formula InChI=1S/C9H16N2O2/c12-9(8-2-1-7-13-8)11-5-3-10-4-6-11/h8,10H,1-7H2
SMILES O=C(C1CCCO1)N2CCNCC2
IUPAC Name oxolan-2-yl(piperazin-1-yl)methanone
Molecular Weight (g/mol) 184.24
Formula Weight 184.1212
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.