missing translation for 'onlineSavingsMsg'
Learn More

1-(Thien-2-ylmethyl)piperidine-4-carboxylic acid hydrochloridehydrate, 97%, Thermo Scientific™

Product Code. 10411934
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10411934 250 mg Amber Glass Bottle 250mg
10205474 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10411934 Supplier Thermo Scientific Maybridge Supplier No. CC66751CB
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 944450-84-4
Molecular Formula C11H18ClNO3S
Molecular Weight (g/mol) 279.779
MDL Number MFCD09817555
InChI Key YUUKLZNSOZECHH-UHFFFAOYSA-N
Synonym 1-thien-2-ylmethyl piperidine-4-carboxylic acid hydrochloride hydrate, 1-thiophen-2-ylmethyl piperidine-4-carboxylic acid hydrate hydrochloride, 1-thiophen-2-ylmethyl piperidine-4-carboxylic acid hydrochloride hydrate, 1-thiophen-2-yl methyl piperidine-4-carboxylic acid-hydrogen chloride-water 1/1/1
PubChem CID 44118590
IUPAC Name 1-(thiophen-2-ylmethyl)piperidine-4-carboxylic acid;hydrate;hydrochloride
SMILES C1CN(CCC1C(=O)O)CC2=CC=CS2.O.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 279.79
Percent Purity 97%
Chemical Name or Material 1-(Thien-2-ylmethyl)piperidine-4-carboxylic acid hydrochloridehydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.