missing translation for 'onlineSavingsMsg'
Learn More

(1-Thieno[3,2-d]pyrimidin-4-ylpiperid-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10285764
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285764 250 mg Amber Glass Bottle 250mg
10718465 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285764 Supplier Thermo Scientific Maybridge Supplier No. CC63813CB
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 937795-94-3
Molecular Formula C12H16N4S
Molecular Weight (g/mol) 248.35
MDL Number MFCD09787494
InChI Key ZKNFKYHZGAWSEG-UHFFFAOYSA-N
Synonym 1-thieno 3,2-d pyrimidin-4-ylpiperid-4-yl methylamine, 1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-thiopheno 2,3-e pyrimidin-4-yl-4-piperidyl methylamine, 4-4-aminomethyl piperidin-1-yl thieno 3,2-d pyrimidine
PubChem CID 24229708
SMILES NCC1CCN(CC1)C1=C2SC=CC2=NC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-(1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanamine
Formula Weight 248.35
Percent Purity 97%
Chemical Name or Material (1-Thieno[3, 2-d]pyrimidin-4-ylpiperid-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.