missing translation for 'onlineSavingsMsg'
Learn More

1-Thioglycerol, TRC

Product Code. 30137983
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137983 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30137983 Supplier Toronto Research Chemicals Supplier No. TRCT344485250G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 96-27-5
Molecular Formula C3 H8 O2 S
Quantity 250 g
Synonym 3-Mercapto-1,2-propanediol, (+/-)-3-Mercapto-1,2-propanediol, 1,2-Dihydroxy-3-mercaptopropane, 1-Mercapto-2,3-dihydroxypropane, 1-Mercapto-2,3-propanediol, 1-Mercaptoglycerol, 1-Monothioglycerol, 1-Thio-2,3-propanediol, 1-Thio-DL-glycerol, 1-Thioglycerol, 2,3-Dihydroxy-1-propanethiol, 2,3-Dihydroxypropanethiol, 3-Mercapto-1,2-dihydroxypropane, DL-1-Thioglycerol, Glycerol-1-thiol, NSC 5370, Thioglycerin, Thioglycerol, Thiovanol, alpha-Monothioglycerol, alpha-Thioglycerin, alpha-Thioglycerol, alpha-Thiolglycerol, (±)-3-Mercapto-1,2-propanediol, α-Monothioglycerol, α-Thioglycerin, α-Thioglycerol, α-Thiolglycero
InChI Formula InChI=1S/C3H8O2S/c4-1-3(5)2-6/h3-6H,1-2H2
SMILES OCC(O)CS
IUPAC Name 3-sulfanylpropane-1,2-diol
Molecular Weight (g/mol) 108.16
Formula Weight 108.0245
Percent Purity >95%
Chemical Name or Material 1-Thioglycerol
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.