missing translation for 'onlineSavingsMsg'
Learn More

1-(trans-2-Phenylethenyl)-4-(phenylethynyl)benzene, 97%

Product Code. 15457775
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
15457775 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 15457775

Brand: Thermo Scientific Alfa Aesar H30638.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21850-30-6 | C22H16 | 280.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21850-30-6
Melting Point 212°C to 214°C
Molecular Formula C22H16
MDL Number MFCD12407077
Quantity 1 g
Synonym 1-trans-2-phenylethenyl-4-phenylethynyl benzene, 1-e-2-phenylethenyl-4-2-phenylethynyl benzene, 4-ethynyl-trans-stilbene, e-4-phenylethynyl stilbene, 4-phenylethynyl-cis-stilbene, 4-trans-beta-styryl diphenylacetylene, 1-phenylethynyl-4-trans-styryl benzene, 1-e-2-phenylethenyl-4-phenylethynyl benzene #
InChI Key UIWXLKXVVDEMKD-UHFFFAOYSA-N
SMILES C(=CC1=CC=C(C=C1)C#CC1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 1-(2-phenylethenyl)-4-(2-phenylethynyl)benzene
Molecular Weight (g/mol) 280.37
PubChem CID 5378113
Formula Weight 280.37
Percent Purity 97%
Chemical Name or Material 1-(trans-2-Phenylethenyl)-4-(phenylethynyl)benzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.