missing translation for 'onlineSavingsMsg'
Learn More

1-(trans-Cinnamoyl)imidazole, 98%, Thermo Scientific Chemicals

Product Code. 11340044
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340044 5 g 5g
11350044 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340044 Supplier Thermo Scientific Alfa Aesar Supplier No. L00441.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1138-15-4
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.225
MDL Number MFCD00005288
InChI Key XVGXMXZUJNAGFZ-VOTSOKGWSA-N
Synonym n-trans-cinnamoylimidazole, trans-cinnamoylimidazole, imidazole, 1-cinnamoyl-, e, imidazole, 1-cinnamoyl-, trans, imidazole, trans, 1-trans-cinnamoyl imidazole, imidazole, e, e-1-1h-imidazol-1-yl-3-phenylprop-2-en-1-one, 1h-imidazole, e, 2e-1-imidazolyl-3-phenylprop-2-en-1-one
PubChem CID 5357650
IUPAC Name (E)-1-imidazol-1-yl-3-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)N2C=CN=C2

Specifications

Melting Point 128°C to 132°C
Odor Odorless
Quantity 5 g
Beilstein 608846
Sensitivity Moisture sensitive
Formula Weight 198.23
Percent Purity 98%
Chemical Name or Material 1-(trans-Cinnamoyl)imidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.