missing translation for 'onlineSavingsMsg'
Learn More

1-Triacontanol, 96%

Product Code. 10154940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10154940 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10154940 Supplier Thermo Scientific Acros Supplier No. 218131000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 593-50-0 | C30H62O | 438.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 593-50-0
CAS Min % 95.0
CAS Max % 100.0
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C30H62O
Linear Formula CH3(CH2)29OH
MDL Number MFCD00002963
Quantity 100 mg
Merck Index 15, 9753
Synonym 1-triacontanol, triacontanol, melissyl alcohol, myricyl alcohol, n-triacontanol, triacontyl alcohol, triacontanol-1, miraculan, myricyl alcohol van, 1-hydroxytriacontane
InChI Key REZQBEBOWJAQKS-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO
IUPAC Name triacontan-1-ol
Molecular Weight (g/mol) 438.82
PubChem CID 68972
ChEBI CHEBI:28409
Formula Weight 438.82
Percent Purity 96%
Physical Form Flakes
Chemical Name or Material 1-Triacontanol, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.