missing translation for 'onlineSavingsMsg'
Learn More

1-Tridecene, 97%

Product Code. 11369849
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369849 25 g 25g
11359849 5 g 5g
11379849 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369849 Supplier Thermo Scientific Alfa Aesar Supplier No. B24413.14
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2437-56-1 | C13H26 | 182.351 g/mol

It is used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2437-56-1
Molecular Formula C13H26
Molecular Weight (g/mol) 182.351
MDL Number MFCD00008976
InChI Key VQOXUMQBYILCKR-UHFFFAOYSA-N
Synonym 1-tridecene, n-tridec-1-ene, tridecylene, alpha-tridecene, undecylethylene, tridecene-1, unii-5b0u2s314c, ccris 5719, 1-tridecylene, .alpha.-tridecene
PubChem CID 17095
IUPAC Name tridec-1-ene
SMILES CCCCCCCCCCCC=C

Especificaciones

Melting Point -23°C
Density 0.766
Boiling Point 232°C to 233°C
Flash Point 79°C (174°F)
Odor Mild
Linear Formula CH3(CH2)10CH=CH2
Refractive Index 1.433
Quantity 25 g
Beilstein 1744660
Solubility Information Insoluble in water.
Formula Weight 182.35
Percent Purity 97%
Chemical Name or Material 1-Tridecene
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.