missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoromethyl)cyclohexane-1-carboxylic acid, Thermo Scientific™

Product Code. 10210060
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10210060 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10210060 Supplier Thermo Scientific Maybridge Supplier No. MAY00136DA
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 180918-40-5
Packaging Amber Glass Bottle
Molecular Formula C8H11F3O2
Quantity 1 g
Synonym 1-trifluoromethyl cyclohexane-1-carboxylic acid, 1-trifluoromethyl cyclohexanecarboxylic acid, cyclohexanecarboxylicacid, 1-trifluoromethyl, acmc-20ap98, 1-trifluoromethyl-cyclohexanecarboxylic acid, cyclohexanecarboxylicacid,1-trifluoromethyl
InChI Key VFSFTDFBHSFDEE-UHFFFAOYSA-N
SMILES C1CCC(CC1)(C(=O)O)C(F)(F)F
IUPAC Name 1-(trifluoromethyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol) 196.169
PubChem CID 15507524
Formula Weight 196.17
Percent Purity 97%
Chemical Name or Material 1-(Trifluoromethyl)cyclohexane-1-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.