missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoromethylcyclopropyl)benzene, 97%, Thermo Scientific™

Product Code. 15493357
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493357 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15493357 Supplier Thermo Scientific Alfa Aesar Supplier No. H26056.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 883547-73-7
Molecular Formula C10H9F3
Refractive Index 1.455
MDL Number MFCD08059543
Quantity 250 mg
UN Number UN1993
Synonym 1-trifluoromethylcyclopropyl benzene, 1-phenyl-1-trifluoromethyl cyclopropane, 1-trifluoromethyl cyclopropyl benzene, trifluoromethyl cyclopropyl benzene, benzene, 1-trifluoromethyl cyclopropyl, 1-trifluoromethyl cycloprop-1-yl benzene
Solubility Information Insoluble in water.
InChI Key RBZAHILDWCBKFX-UHFFFAOYSA-N
SMILES C1CC1(C2=CC=CC=C2)C(F)(F)F
IUPAC Name [1-(trifluoromethyl)cyclopropyl]benzene
Molecular Weight (g/mol) 186.177
PubChem CID 20405523
Formula Weight 186.18
Percent Purity 97%
Chemical Name or Material 1-(Trifluoromethylcyclopropyl)benzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.