missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsilyl)imidazole, 97%

Product Code. 11498710
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
250 g
missing translation for 'unitSize':
100g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity missing translation for 'unitSize'
11498710 100 g 100g
11488710 5 g 5g
11444164 50 g 50g
11454164 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 11498710 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A12512.22
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 18156-74-6 | C6H12N2Si | 140.26 g/mol

1-(Trimethylsilyl)imidazole acts as a silylating agent for alcohols, carbohydrates and 1,3-dicarbonyl compounds. It is a derivatization reagent used for selective silylation of hydroxyl groups and carboxylic acid without affecting the amino group. It employed as an intermediate for the synthesis of imidazole derivatives. It plays an essential role to synthesize polysubstituted chiral spirotetrahydropyrans. It is also used in the quantification of pintol after derivatization by gas chromatography/ mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trimethylsilyl)imidazole acts as a silylating agent for alcohols, carbohydrates and 1,3-dicarbonyl compounds. It is a derivatization reagent used for selective silylation of hydroxyl groups and carboxylic acid without affecting the amino group. It employed as an intermediate for the synthesis of imidazole derivatives. It plays an essential role to synthesize polysubstituted chiral spirotetrahydropyrans. It is also used in the quantification of pintol after derivatization by gas chromatography/ mass spectrometry.

Solubility
Miscible with most common organic solvents. It decomposes in water.

Notes
Moisture and light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name imidazol-1-yl(trimethyl)silane
SMILES C[Si](C)(C)N1C=CN=C1

Spécification

Melting Point -42°C
Density 0.956
Boiling Point 212°C
Flash Point 18°C (64°F)
Odor Weak
Refractive Index 1.4765
Quantity 100 g
UN Number UN1993
Beilstein 606148
Sensitivity Moisture sensitive
Solubility Information Miscible with most common organic solvents. It decomposes in water.
Formula Weight 140.26
Percent Purity 97%
Chemical Name or Material 1-(Trimethylsilyl)imidazole
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.