missing translation for 'onlineSavingsMsg'
Learn More

1-Undecene, 97%

Product Code. 15400158
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400158 5 g 5g
15410158 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400158 Supplier Thermo Scientific Alfa Aesar Supplier No. H53387.06
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 821-95-4 | C11H22 | 154.297 g/mol

1-Undecene was used as a biomarker for identification of Pseudomonas aeruginosa strain. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Undecene was used as a biomarker for identification of Pseudomonas aeruginosa strain. It is also used as pharmaceutical intermediate.

Solubility
Solunle in ether, chloroform and ligroin. Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-95-4
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
MDL Number MFCD00008956
InChI Key DCTOHCCUXLBQMS-UHFFFAOYSA-N
Synonym 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720
PubChem CID 13190
ChEBI CHEBI:77444
IUPAC Name undec-1-ene
SMILES CCCCCCCCCC=C

Specifications

Melting Point -49°C
Density 0.751
Boiling Point 192°C to 193°C
Flash Point 63°C (145°F)
Linear Formula CH3(CH2)8CH=CH2
Refractive Index 1.426
Quantity 5 g
Beilstein 1740044
Solubility Information Solunle in ether,chloroform and ligroin. Insoluble in water.
Formula Weight 154.3
Percent Purity 97%
Chemical Name or Material 1-Undecene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Undecene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Undecene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.