missing translation for 'onlineSavingsMsg'
Learn More

1-Undecyne, 97%

Product Code. 15438809
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438809 2 g 2g
11337885 10 g 10g
11347885 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15438809 Supplier Thermo Scientific Alfa Aesar Supplier No. L02883.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2243-98-3 | C11H20 | 152.281 g/mol

1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

Solubility
Miscible with acetone, ether, ethanol and benzene. Slightly miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2243-98-3
Molecular Formula C11H20
MDL Number MFCD00015063
InChI Key YVSFLVNWJIEJRV-UHFFFAOYSA-N
PubChem CID 75249
ChEBI CHEBI:87545
IUPAC Name undec-1-yne
SMILES CCCCCCCCCC#C

Specifications

Melting Point -25°C
Density 0.77
Boiling Point 196°C
Flash Point 65°C
Odor Characteristic
Linear Formula CH3(CH2)8C≡CH
Refractive Index 1.431
Quantity 2 g
Beilstein 1740043
Synonym 1-undecyne, rutylidene, unii-y07u3q24us, undecyne, acmc-1 cr7r, 1-undecyne 10 g
Solubility Information Miscible with acetone,ether,ethanol and benzene. Slightly miscible with water.
Molecular Weight (g/mol) 152.28
Formula Weight 152.28
Percent Purity 97%
Chemical Name or Material 1-Undecyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.