missing translation for 'onlineSavingsMsg'
Learn More

1-Undecyne, 97%

Product Code. p-7043817
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 15438809

Brand: Thermo Scientific Alfa Aesar L02883.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2243-98-3 | C11H20 | 152.281 g/mol

1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

Solubility
Miscible with acetone, ether, ethanol and benzene. Slightly miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2243-98-3
Molecular Formula C11H20
MDL Number MFCD00015063
InChI Key YVSFLVNWJIEJRV-UHFFFAOYSA-N
PubChem CID 75249
ChEBI CHEBI:87545
IUPAC Name undec-1-yne
SMILES CCCCCCCCCC#C

Specifications

Melting Point -25°C
Density 0.77
Boiling Point 196°C
Flash Point 65°C
Odor Characteristic
Linear Formula CH3(CH2)8C≡CH
Refractive Index 1.431
Quantity 2 g
Beilstein 1740043
Synonym 1-undecyne, rutylidene, unii-y07u3q24us, undecyne, acmc-1 cr7r, 1-undecyne 10 g
Solubility Information Miscible with acetone,ether,ethanol and benzene. Slightly miscible with water.
Molecular Weight (g/mol) 152.28
Formula Weight 152.28
Percent Purity 97%
Chemical Name or Material 1-Undecyne
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.