missing translation for 'onlineSavingsMsg'
Learn More

1-Vinyl-2-pyrrolidinone, TRC

Product Code. 17970405
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970405 50 g 50g
17953265 10 g 10g
17951785 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970405 Supplier Toronto Research Chemicals Supplier No. TRCV49990050G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-12-0
Molecular Formula C6 H9 N O
Molecular Weight (g/mol) 111.14
Synonym 1-Ethenylpyrrolidin-2-one, N-Vinyl-2-pyrrolidone, Povidone Imp. A (EP), 1-Vinylpyrrolidin-2-one, 1-Ethenyl-2-pyrrolidinone, 1-Vinyl-2-pyrrolidinone, 1-Vinyl-2-pyrrolidone, Aronix M 150, N-V 2RC, N-VP, N-Vinyl-2-pyrrolidinone, N-Vinylpyrrolin-2-one, NSC 10222, NSC 683040, NVP 300, V-Pyrol, V-Pyrol RC, Vinylbutyrolactam
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES C=CN1CCCC1=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2
Formula Weight 111.0684
Percent Purity >95%
Chemical Name or Material N-Vinyl-2-pyrrolidone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.