missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylimidazole, 99%

Product Code. 11312917
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312917 100 g 100g
11302917 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312917 Supplier Thermo Scientific Alfa Aesar Supplier No. L16174.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1072-63-5 | C5H6N2 | 94.12 g/mol

1-Vinylimidazole is used as a monomer in the synthesis of molecularly imprinted polymers. It is involved in the removal of vinyl protecting groups with an isoprene-catalyzed lithiation process. Also, it is involved in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) microbeads. Further, it is used to prepare ion imprinted polyvinylimidazole-silica hybrid copolymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Vinylimidazole is used as a monomer in the synthesis of molecularly imprinted polymers. It is involved in the removal of vinyl protecting groups with an isoprene-catalyzed lithiation process. Also, it is involved in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) microbeads. Further, it is used to prepare ion imprinted polyvinylimidazole-silica hybrid copolymer.

Solubility
Miscible with water.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-63-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00005297
InChI Key OSSNTDFYBPYIEC-UHFFFAOYSA-N
Synonym 1-vinylimidazole, 1-vinyl-1h-imidazole, n-vinylimidazole, 1h-imidazole, 1-ethenyl, lufixan, polyvinylimidazole, poly vinylimidazole, poly n-vinylimidazole, poly 1-vinylimidazole, n-vinylimidazole polymer
PubChem CID 66171
SMILES C=CN1C=CN=C1

Specifications

Melting Point <-50°C
Density 1.038
Boiling Point 192°C to 194°C
Flash Point 84°C (183°F)
Refractive Index 1.5315
Quantity 100 g
UN Number UN3267
Beilstein 105321
Sensitivity Hygroscopic
Solubility Information Miscible with water.
IUPAC Name 1-ethenyl-1H-imidazole
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 1-Vinylimidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.