missing translation for 'onlineSavingsMsg'
Learn More

10-(1-Naphthyl)anthracene-9-boronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15469617
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469617 250 mg 250mg
15459617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469617 Supplier Thermo Scientific Alfa Aesar Supplier No. H52842.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400607-46-7
Molecular Formula C24H17BO2
Molecular Weight (g/mol) 348.21
MDL Number MFCD11977302
InChI Key ASQXKNXJNDLXQV-UHFFFAOYSA-N
Synonym 10-naphthalen-1-yl anthracen-9-yl boronic acid, 10-1-naphthalenyl-9-anthracenyl boronic acid, 10-naphthalene-1-yl-9-anthracene boronic acid, 10-1-naphthyl anthracene-9-boronic acid, 10-naphthalen-1-yl anthracen-9-ylboronic acid, 10-napthalene-1-yl-9-anthracene boronic acid, 10-1-naphthyl-9-anthryl boronic acid, 10-1-naphthyl anthracene-9-ylboronic acid
PubChem CID 23088558
IUPAC Name (10-naphthalen-1-ylanthracen-9-yl)boronic acid
SMILES OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1

Specifications

Quantity 250 mg
Formula Weight 348.21
Percent Purity 97%
Chemical Name or Material 10-(1-Naphthyl)anthracene-9-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.