missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydro-5H-dibenzo[b,f]azepine, TRC

Product Code. 30136667
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136667 25 g 25g
30133971 100 g 100g
17983745 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136667 Supplier Toronto Research Chemicals Supplier No. TRCD44887025G
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494-19-9
Molecular Formula C14 H13 N
Molecular Weight (g/mol) 195.26
Synonym Iminodibenzyl, 10,11-Dihydro-5H-dibenzo[b,f]azepine, Clomipramine Hydrochloride Imp. E (EP), Trimipramine Maleate Imp. F (EP), Carbamazepine Imp. E (EP)
IUPAC Name 6,11-dihydro-5H-benzo[b][1]benzazepine
SMILES C1Cc2ccccc2Nc3ccccc13

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H13N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-8,15H,9-10H2
Formula Weight 195.1048
Chemical Name or Material Iminodibenzyl (10,11-Dihydro-5H-dibenzo[b,f]azepine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.