missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydro Carbamazepine, TRC

Product Code. 30379530
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379530 1 g
30384855 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379530 Supplier Toronto Research Chemicals Supplier No. TRCD4487651G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3564-73-6
Molecular Formula C15 H14 N2 O
Molecular Weight (g/mol) 238.28
Synonym 10,11-Dihydro-5H-dibenzo[b,f]-azepine-5-carboxamide, Carbamazepine Imp. A (EP), 10,11-Dihydro-carbamazepine, Carbamazepine-10,11-dihydro
IUPAC Name 5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
SMILES NC(=O)N1c2ccccc2CCc3ccccc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H14N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-8H,9-10H2,(H2,16,18)
Formula Weight 238.1106
Chemical Name or Material 10,11-Dihydro-carbamazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.