missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydrocarbamazepine, 99%

Product Code. 11314016
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314016 50 mg 50mg
11324016 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314016 Supplier Thermo Scientific Alfa Aesar Supplier No. L17271.MF
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3564-73-6 | C15H14N2O | 238.29 g/mol

10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.

Solubility
Soluble in dichloromethane. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3564-73-6
Molecular Formula C15H14N2O
Molecular Weight (g/mol) 238.29
MDL Number MFCD00005072
InChI Key PHNLCHMJDSSPDQ-UHFFFAOYSA-N
Synonym 10,11-dihydrocarbamazepine, 10,11-dihydro-5h-dibenzo b,f azepine-5-carboxamide, unii-ec017va9fr, dihydrocarbamazepine, ec017va9fr, 5h-dibenz b,f azepine-5-carboxamide, 10,11-dihydro, chembl41543, 5,6-dihydrobenzo b 1 benzazepine-11-carboxamide, 10,11-dihydro-5h-dibenz b,f azepine-5-carboxamide
PubChem CID 19099
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-2-carboxamide
SMILES NC(=O)N1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

Melting Point 204°C to 206°C
Quantity 50 mg
Beilstein 196969
Solubility Information Soluble in dichloromethane. Insoluble in water.
Formula Weight 238.29
Percent Purity 99%
Chemical Name or Material 10,11-Dihydrocarbamazepine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.