missing translation for 'onlineSavingsMsg'
Learn More

10,12-Tricosadiynoic acid, 96%

Product Code. 11398305
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11398305

Brand: Thermo Scientific Alfa Aesar L10231.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66990-30-5 | C23H38O2 | 346.555 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66990-30-5
Molecular Formula C23H38O2
Molecular Weight (g/mol) 346.555
MDL Number MFCD00041683
InChI Key DIEDVCMBPCRJFQ-UHFFFAOYSA-N
Synonym 10,12-tricosadiynoic acid, 10,12-tricosadiynoicacid, acmc-209nx8, 10,12-tricosadiynoic acid gc
PubChem CID 538457
IUPAC Name tricosa-10,12-diynoic acid
SMILES CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O

Specifications

Melting Point 55°C to 59°C
Quantity 1 g
Beilstein 1880414
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 346.56
Percent Purity 96%
Chemical Name or Material 10,12-Tricosadiynoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.