missing translation for 'onlineSavingsMsg'
Learn More

10-Acetoxy-5H-dibenz[b,f]azepine-5-carboxamide, TRC

Product Code. 30138253
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138253 100 mg 100mg
30006926 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138253 Supplier Toronto Research Chemicals Supplier No. TRCA165050100MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 952740-00-0
Molecular Formula C17 H14 N2 O3
Molecular Weight (g/mol) 294.3
Synonym 10-(Acetyloxy)-5H-dibenz[b,f]azepine-5-carboxamide
IUPAC Name 10-Acetoxy-5H-dibenz[b,f]azepine-5-carboxamide
SMILES CC(=O)OC1=Cc2ccccc2N(C(=O)N)c3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H14N2O3/c1-11(20)22-16-10-12-6-2-4-8-14(12)19(17(18)21)15-9-5-3-7-13(15)16/h2-10H,1H3,(H2,18,21)
Formula Weight 294.1004
Chemical Name or Material 10-Acetoxy-5H-dibenz[b,f]azepine-5-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.