missing translation for 'onlineSavingsMsg'
Learn More

10-Deacetyl-7,10-dimethoxy-Baccatin III, TRC

Product Code. 30135486
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135486 100 mg 100mg
17972935 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135486 Supplier Toronto Research Chemicals Supplier No. TRCD198890100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183133-94-0
Molecular Formula C31 H40 O10
Molecular Weight (g/mol) 572.64
Synonym (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(Acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-9,11-dihydroxy-4,6-dimethoxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, [2aR-(2aα,4β,4aβ,6β,9α,11α,12α,12aα,12bα)]-12b-(Acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-9,11-dihydroxy-4,6-dimethoxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, CBT-1A
IUPAC Name [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4-acetyloxy-1,15-dihydroxy-9,12-dimethoxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
SMILES CO[C@H]1C[C@H]2OC[C@@]2(OC(=O)C)[C@H]3[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](O)C(=C([C@@H](OC)C(=O)[C@]13C)C5(C)C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C31H40O10/c1-16-19(33)14-31(36)26(40-27(35)18-11-9-8-10-12-18)24-29(5,25(34)23(38-7)22(16)28(31,3)4)20(37-6)13-21-30(24,15-39-21)41-17(2)32/h8-12,19-21,23-24,26,33,36H,13-15H2,1-7H3/t19-,20-,21+,23+,24-,26-,29+,30-,31+/m0/s1
Formula Weight 572.2621
Chemical Name or Material 10-Deacetyl-7,10-dimethoxy-Baccatin III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.