missing translation for 'onlineSavingsMsg'
Learn More

10-Deoxo-9,10-dehydro Ketotifen, TRC

Product Code. 30135074
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135074 50 mg 50mg
30011358 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135074 Supplier Toronto Research Chemicals Supplier No. TRCD23158050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4673-38-5
Molecular Formula C19 H19 N S
Molecular Weight (g/mol) 293.43
Synonym Piperidine, 4-(4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methyl-, 4-(4H-Benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine, 4H-Benzo[4,5]cyclohepta[1,2-b]thiophene, piperidine deriv., 4-(4H-Benzo[4,5]cyclohepta[1,2-b]thiophen-4-ylidene)-1-methylpiperidine, Ketotifen Hydrogen Fumarate Imp. A (EP)
IUPAC Name 4-benzo[1,2]cyclohepta[2,4-b]thiophen-10-ylidene-1-methylpiperidine
SMILES CN1CCC(=C2c3ccccc3C=Cc4sccc24)CC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H19NS/c1-20-11-8-15(9-12-20)19-16-5-3-2-4-14(16)6-7-18-17(19)10-13-21-18/h2-7,10,13H,8-9,11-12H2,1H3
Formula Weight 293.1238
Chemical Name or Material 10-Deoxo-9,10-dehydro Ketotifen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.