missing translation for 'onlineSavingsMsg'
Learn More

10-Methylphenothiazine, 98%

Product Code. 11490207
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490207 5 g 5g
11400217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490207 Supplier Thermo Scientific Alfa Aesar Supplier No. B21648.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1207-72-3 | C13H11NS | 213.30 g/mol

They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207-72-3
Molecular Formula C13H11NS
Molecular Weight (g/mol) 213.30
MDL Number MFCD00041836
InChI Key QXBUYALKJGBACG-UHFFFAOYSA-N
Synonym 10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
PubChem CID 71015
IUPAC Name 10-methylphenothiazine
SMILES CN1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 100°C to 103°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 213.3
Percent Purity 98%
Chemical Name or Material 10-Methylphenothiazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.