missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CBR-470-1
GREENER_CHOICE

Product Code. 18788956
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18788956

Brand: Tocris Bioscience™ 7018/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PGK1 inhibitior; activates Nrf2

CBR-470-1 is a phosphoglycerate kinase 1 (PGK1) inhibitor. CBR-470-1 promotes accumulation of the reactive metabolites methylglyoxal (MGO) resulting in KEAP1 modification and dimerization, which in turn leads to activation of the Nrf2 transcriptional program. CBR-470-1 protects SH-SY5Y neuroblastoma cells from MPP+-induced cytotoxicity via the Keap1-Nrf2 cascade.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 R,4 S)-rel-4-[(4-Chlorophenyl)sulfonyl]tetrahydro-N-(2-methylpropyl)-3-thiophenamine-1,1-dioxide
CAS 2416095-06-0
Quantity 10 mg
Target Nrf2 Activators
Molecular Formula C14H20ClNO4S2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.