missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HIOC, >98%
GREENER_CHOICE

Product Code. 30000834
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000834

Brand: Tocris Bioscience™ 5961/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TrkB receptor agonist; neuroprotectant

Selective TrkB receptor agonist. Protects neurons from excitotoxicity. Protects retinas from light-induced retinal degeneration (LIRD) in vivo. Blood brain barrier and blood-retinal barrier penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 314054-36-9
Molecular Formula C16H19N3O3

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 301.34
Purity >98%
Molecular Weight (g/mol) 301.346
Chemical Name or Material N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]-2-oxo-3-piperidinecarboxamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.