missing translation for 'onlineSavingsMsg'
Learn More

10-Oxo Docetaxel, MedChemExpress

Product Code. 16479657
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479657 5 mg 5mg
16477118 10 mg 10mg
16441629 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16479657 Supplier Medchem Express Supplier No. HY16674
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 10-Oxo Docetaxel (Docetaxel Impurity 1) is a novel taxoid having remarkable anti-tumor properties and a Docetaxel intermediate.

10-Oxo Docetaxel (Docetaxel Impurity 1) is a novel taxoid having remarkable anti-tumor properties and a Docetaxel intermediate.

Specifications

Chemical Name or Material 10-Oxo Docetaxel
CAS 167074-97-7
Color White
Purity Grade Notes Research
Molecular Formula C43H51NO14
Quantity 5 mg
Synonym Docetaxel Impurity 1
SMILES CC(O[C@@]1([C@](C[C@@H]2O)([H])OC1)[C@@]([C@@]2(C3=O)C)([H])[C@@H]([C@@](C4(C)C)(C[C@H](OC([C@H](O)[C@H](C5=CC=CC=C5)NC(OC(C)(C)C)=O)=O)C(C)=C4C3=O)O)OC(C6=CC=CC=C6)=O)=O
Molecular Weight (g/mol) 805.86
Formula Weight 805.86
Percent Purity 93.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.