missing translation for 'onlineSavingsMsg'
Learn More

10-Undecen-1-ol, 99%

Product Code. 11487856
Click to view available options
Quantity:
25 g
100 g
500 g
Förpackningsstorlek
100g
25g
500g
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11487856

missing translation for 'mfr': Thermo Scientific Alfa Aesar A14002.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 112-43-6 | C11H22O | 170.296 g/mol

10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 112-43-6
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00004750
InChI Key GIEMHYCMBGELGY-UHFFFAOYSA-N
Synonym 10-undecen-1-ol, undecylenic alcohol, undecylenyl alcohol, 10-undecenol, 1-undecen-11-ol, 10-undecylen-1-ol, 11-hydroxy-1-undecene, omega-undecenyl alcohol, undecen-1-ol, .omega.-undecenyl alcohol
PubChem CID 8185
IUPAC Name undec-10-en-1-ol
SMILES C=CCCCCCCCCCO

Specifikationer

Melting Point -3°C
Density 0.848
Boiling Point 132°C to 133°C (15 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.45
Quantity 500 g
Beilstein 1753225
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 170.3
Percent Purity 99%
Chemical Name or Material 10-Undecen-1-ol
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.