missing translation for 'onlineSavingsMsg'
Learn More

10-Undecynoic Acid, TRC

Product Code. 30451989
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451989 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451989 Supplier Toronto Research Chemicals Supplier No. TRCU789030100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Undecynoic Acid
CAS 2777-65-3
Molecular Formula C11H18O2
Quantity 100 mg
Synonym 10-Hendecynoic Acid
InChI Formula InChI=1S/C11H18O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h1H,3-10H2,(H,12,13)
SMILES OC(=O)CCCCCCCCC#C
IUPAC Name undec-10-ynoic acid
Molecular Weight (g/mol) 182.26
Formula Weight 182.1307
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.