missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ 100% Recycled Polyethylene Ramp for Square EcoPolyBlend™ Spill Control Pallet

Product Code. 12941415
Change view
Click to view available options
Material:
Recycled Polyethylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
12941415 Recycled Polyethylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12941415 Supplier Justrite™ Supplier No. 28688

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Increase workplace safety and improve cleaniliness while using environmentally friendly products. The Justrite™ 100% Recycled Polyethylene Ramp for Square EcoPolyBlend™ Spill Control Pallet is an accessory to help facilitate the use of spill control pallets.

Ramp for 4-drum square pallets clips securely onto pallet and offers convenience when loading and unloading heavy drums.

Spécification

Color Black
Material Recycled Polyethylene
Dimensions (L x W x H) 33 x 10.25 x 49 in.
Width (English) 49 in.
Height (English) 10.25 in.
Depth (English) 33 in.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.