missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NLS-StAx-h

Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
NLS-StAx-h is a Wnt inhibitor (IC50 = 1.4 μM). Stapled peptide inhibitor of oncogenic Wnt signaling. Inhibits β-catenin-transcription factor interactions. Inhibits proliferation and migration of colorectal cancer cells. Cell permeable.
Specifications
Specifications
| CAS | 2872559-21-0 |
| Content And Storage | Store at -20°C |
| Quantity | 100 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction