Learn More
Hamilton™ 1000 Series Standard PTFE Instrument Syringe
Setting the standard for quality and reliability.
161.00 € - 455.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10132963
|
Hamilton™
5495-55 |
12.5 mL |
455.00 €
Each |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11560222
|
Hamilton™
5495-45 |
2.5 mL |
161.00 €
Each |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Syringe life is exceptional when used in conjunction with Hamilton™ precision syringe drives
- Enables precision liquid handling
- Gastight for leak-free performance