missing translation for 'onlineSavingsMsg'
Learn More

10058-F4

Product Code. 15447019
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447019 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447019 Supplier Thermo Scientific Alfa Aesar Supplier No. J65650.MA
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 403811-55-2 | C12H11NOS2 | 249.346 g/mol

A c-Myc inhibitor that induces apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 403811-55-2
Color Yellow
Molecular Formula C12H11NOS2
MDL Number MFCD00758517
Quantity 10 mg
Synonym c-myc inhibitor, 5e-5-4-ethylbenzylidene-2-mercapto-1,3-thiazol-4 5h-one, 5-4-ethylphenyl methylene-2-thioxo-4-thiazolidinone, e-5-4-ethylbenzylidene-2-mercaptothiazol-4 5h-one, z,e-5-4-ethylbenzylidine-2-thioxothiazolidin-4-one, 5e-5-4-ethylphenyl methylidene-2-sulfanyl-1,3-thiazol-4-one, 5e-5-4-ethylphenyl methylidene-2-sulfanylidene-1,3-thiazolidin-4-one, 10058-f hplc , solid
Solubility Information Soluble in DMSO; Soluble in water at concentrations less than 2mg/ml
InChI Key SVXDHPADAXBMFB-JXMROGBWSA-N
SMILES CCC1=CC=C(C=C1)C=C2C(=O)NC(=S)S2
IUPAC Name (5E)-5-[(4-ethylphenyl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol) 249.346
PubChem CID 1271002
Formula Weight 249.35
Physical Form Solid
Chemical Name or Material 10058-F4
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.