missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Cr/V, AISI E6150, G61500, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, Cr/V, AISI E6150, G61500, 100g
Specifications
Specifications
| Chemical Name or Material | Niobium/Titanium/Nitrogen/Cobalt/Tin/Phosphorus/Aluminium/Molybdenum/Nickel/Vanadium/Copper/Silicon/Carbon/Manganese/Chromium/Boron/Calcium/Oxygen/Zirconium/Arsenic/Tungsten |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Niobium 0.003 %/Titanium 0.003 %/Nitrogen 0.0076 %/Cobalt 0.009 %/Tin 0.009 %/Phosphorus 0.01 %/Aluminium 0.014 %/Molybdenum 0.044 %/Nickel 0.14 %/Vanadium 0.17 %/Copper 0.18 %/Silicon 0.22 %/Carbon 0.51 %/Manganese 0.78 %/Chromium 0.99 %/Boron (0.0002) %/Calcium (0.0002) %/Oxygen (0.0014) %/Zirconium (0.002) %/Arsenic (0.006) %/Tungsten (0.016) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction